Merck
Enzymatic synthesis of bile acid derivatives and biological evaluation against Trypanosoma cruzi.
Bioorganic & medicinal chemistry (2015-06-15)
Guadalupe García Liñares, M Antonela Zígolo, Leandro Simonetti, Silvia A Longhi, Alicia Baldessari
ABSTRAKT
Enzyme catalysis was applied to synthesize derivatives of three bile acids and their biological activity was evaluated as growth inhibitors of the protozoan Trypanosoma cruzi. Twelve mono-, diacetyl and ester derivatives of deoxycholic, chenodeoxycholic and lithocholic acid, seven of them new compounds, were obtained through lipase-catalyzed acetylation, esterification and alcoholysis reactions in very good to excellent yield and a highly regioselective way. Among them, acetylated ester products, in which the lipase catalyzed both reactions in one-pot, were obtained. The influence of various reaction parameters in the enzymatic reactions, such as enzyme source, acylating agent/substrate ratio, enzyme/substrate ratio, solvent and temperature, was studied. Some of the evaluated compounds showed a remarkable activity as Trypanosoma cruzi growth inhibitors, obtaining the best results with ethyl chenodeoxycholate 3-acetate and chenodeoxycholic acid 3,7-diacetate, which showed IC50: 8.6 and 22.8 μM, respectively. In addition, in order to shed light to bile acids behavior in enzymatic reactions, molecular modeling was applied to some derivatives. The advantages showed by the enzymatic methodology, such as mild reaction conditions and low environmental impact, make the biocatalysis a convenient way to synthesize these bile acid derivatives with application as potential antiparasitic agents.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS