Merck
Engineered biofiltration for the removal of disinfection by-product precursors and genotoxicity.
Water research (2015-06-13)
Michael J McKie, Liz Taylor-Edmonds, Susan A Andrews, Robert C Andrews
ABSTRAKT
Disinfection by-products (DBPs) are formed when naturally occurring organic matter reacts with chlorine used in drinking water treatment, and DBPs formed in chlorinated drinking water samples have been shown to cause a genotoxic response. The objective of the current study was to further understand the principles of biofiltration and the resulting impacts on the formation of DBPs and genotoxicity. Pilot-scale systems were utilized to assess the performance of engineered biofilters enhanced with hydrogen peroxide, in-line coagulants, and nutrients when compared to passively operated biofilters and conventional treatment (coagulation, flocculation, sedimentation, non-biological filtration). Organic fractionation was completed using liquid chromatography-organic carbon detection (LC-OCD). Water samples were chlorinated after collection and examined for the removal of trihalomethane (THM), haloacetic acid (HAA), and adsorbable organic halide (AOX) precursors. Additionally, the formation potential of two halogenated furanones, 3-chloro-4(dichloromethyl)-2(5H)-furanone (MX) and mucochloric acid (MCA), and genotoxicity was determined. Biofiltration was shown to preferentially remove more DBP precursors than dissolved organic carbon (DOC). Formation potential of the unregulated DBPs, including MX and MCA, and genotoxic response was shown to be correlated to THM formation. These results infer that monitoring for THMs and HAAs provide insight to the formation of more mutagenic DBPs such as halogenated furanones, and that biofiltration may preferentially remove precursors to DBPs at a rate exceeding the removal of DOC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%