Merck
  • Comparative analysis of Rb1, P16 and ER as diagnostic, prognostic and potential targets for therapeutic agents in ovarian epithelial tumors: an immunohistochemical study of 130 ovarian carcinomas.
Comparative analysis of Rb1, P16 and ER as diagnostic, prognostic and potential targets for therapeutic agents in ovarian epithelial tumors: an immunohistochemical study of 130 ovarian carcinomas.
Journal of ovarian research (2015-06-06)
Donna Catherine Ferguson, Daniel Jerad Long, Megan Christine Smith, Laura Deeanne Craig-Owens, Julie Means, Oluwole Fadare, Mohamed Mokhtar Desouki
ABSTRAKT
Deregulation of CDK4/6, cyclin D/P16 and retinoblastoma (Rb) are known aberrations in certain malignancies. There has been a recent interest in exploring the combination of letrozole and CDK4/6 inhibitors in recurrent ER+ ovarian cancers. This study aimed to determine the frequency of expression of Rb1, P16 and ER in ovarian epithelial tumors by immunohistochemistry. Co-expression of all 3 markers studied was seen in 10% of high grade serous carcinoma (HGSC) and low grade serous carcinoma (LGSC). Coordinate expression of Rb1+ and ER+ in HGSC and LGSC was seen in 67% of grade 1/2 vs. 44 % of grade three tumors (p < 0.05). The reverse was true with positive P16 staining in 73% of grade three vs. 32% of grade 1/2 tumors (p < 0.001). Coordinate pattern of Rb1+ and ER+ in HGSC and LGSC is 19 and 50%, respectively. Rb1 and P16 show inverse expression pattern according to tumor grade with more frequent Rb1 in low grade vs. more frequent P16 in grade 3 tumors. These data provide a rational basis for clinical trials that aim to target these proteins.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Trimethylaluminum, 97%