Merck
New pregnane glycoside derivative from Caralluma retrospiciens (Ehrenb).
Natural product research (2015-01-24)
Mahmoud Fahmi Elsebai, Ietidal El-Tahir Mohamed
ABSTRAKT
Retrospinoside (1) is a new polyoxy pregnane glycoside which was isolated and characterised from the aerial parts of Caralluma retrospiciens (Ehrenb.) N. E. Br., family Apocynaceae. The structure was established as 3-O-[β-D-glucopyranosyl-(1 → 4)-β-D-(3-O-methyl-6-desoxygalactopyranosy)]-14,15,20-trihydroxy-4β-pregnane. Its structural elucidation was performed through extensive spectroscopic measurements including 1D- and 2D-NMR, and HRMS, in addition to chemical methods.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS