Merck
Evolution of the magnetic structure with chemical composition in spinel iron oxide nanoparticles.
Nanoscale (2015-07-24)
G Muscas, N Yaacoub, G Concas, F Sayed, R Sayed Hassan, J M Greneche, C Cannas, A Musinu, V Foglietti, S Casciardi, C Sangregorio, D Peddis
ABSTRAKT
Magnetic properties of iron oxide nanoparticles with spinel structure are strictly related to a complex interplay between cationic distribution and the presence of a non-collinear spin structure (spin canting). With the aim to gain better insight into the effect of the magnetic structure on magnetic properties, in this paper we investigated a family of small crystalline ferrite nanoparticles of the formula CoxNi1-xFe2O4 (0 ≤x≤ 1) having equal size (≈4.5 nm) and spherical-like shape. The field dependence of magnetization at low temperatures indicated a clear increase of magnetocrystalline anisotropy and saturation magnetization (higher than the bulk value for CoFe2O4: ∼130 A m(2) kg(-1)) with the increase of cobalt content. The magnetic structure of nanoparticles has been investigated by Mössbauer spectroscopy under an intense magnetic field (8 T) at a low temperature (10 K). The magnetic properties have been explained in terms of an evolution of the magnetic structure with the increase of cobalt content. In addition a direct correlation between cationic distribution and spin canting has been proposed, explaining the presence of a noncollinear spin structure in terms of superexchange interaction energy produced by the average cationic distribution and vacancies in the spinel structure.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triethylene glycol, BioUltra, anhydrous, ≥99.0% (GC)
Sigma-Aldrich
Triethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, 99.999% trace metals basis