Merck
Synthesis, carbonic anhydrase inhibition and cytotoxic activity of novel chromone-based sulfonamide derivatives.
European journal of medicinal chemistry (2015-04-29)
Fadi M Awadallah, Tamer A El-Waei, Mona M Hanna, Safinaz E Abbas, Mariangela Ceruso, Beyza Ecem Oz, Ozen Ozensoy Guler, Claudiu T Supuran
ABSTRAKT
Four series of sulfonamides incorporating chromone moieties were synthesized and assessed for their cytotoxic activity against MCF-7 and A-549 cell lines, considering the fact that some of these tumors overexpress isoforms of carbonic anhydrase (CA, EC 4.2.1.1) which is inhibited by sulfonamides. Most new sulfonamides showed weak inhibitory activity against the offtarget, cytosolic isoforms hCA I, II but effectively inhibited the tumor-associated hCA IX and XII. The most active compounds featured a primary SO2NH2 group and were active in the low micromolar range against MCF-7 and A-549 cell lines. Compound 4a showed IC50 of 0.72 and 0.50 μM against MCF-7 and A-549 cell lines, respectively, and was further evaluated for its proapoptotic activity which proved enhanced in both tumor types.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Dihydrocoumarin, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Dihydrocoumarin, ≥99%, FCC, FG
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%