Merck
Calendering as a direct shaping tool for the continuous production of fixed-dose combination products via co-extrusion.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-08-02)
A-K Vynckier, H Lin, J A Zeitler, J-F Willart, E Bongaers, J Voorspoels, J P Remon, C Vervaet
ABSTRAKT
In this study calendering is used as a downstream technique to shape monolithic co-extruded fixed-dose combination products in a continuous way. Co-extrudates with a metoprolol tartrate-loaded sustained-release core and a hydrochlorothiazide-loaded immediate-release coat were produced and immediately shaped into a monolithic drug delivery system via calendering, using chilled rolls with tablet-shaped cavities. In vitro metoprolol tartrate release from the ethylcellulose core of the calendered tablets was prolonged in comparison with the sustained release of a multiparticulate dosage form, prepared manually by cutting co-extrudates into mini-matrices. Analysis of the dosage forms using X-ray micro-computed tomography only detected small differences between the pore structure of the core of the calendered tablet and the mini-matrices. Diffusion path length was shown to be the main mechanism behind the release kinetics. Terahertz pulsed imaging visualized that adhesion between the core and coat of the calendered tablet was not complete and a gradient in coat thickness (varying from 200 to 600μm) was observed. Modulated differential scanning calorimetry and X-ray diffraction indicated that the solid-state properties of both drugs were not affected by the calendering procedure.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Dibutyl sebacate, ≥97.0% (GC)