Merck
Growth factor therapy and lymph node graft for lymphedema.
The Journal of surgical research (2015-03-18)
Tomi V Tervala, Pauliina Hartiala, Tuomas Tammela, Mikko T Visuri, Seppo Ylä-Herttuala, Kari Alitalo, Anne M Saarikko
ABSTRAKT
Lymphedema still remains an unsolved problem. Secondary lymphedema often develops after cancer operations or radiation therapy, especially in breast cancer patients. Using a mouse model, we show here that the lymphatic network can be regenerated using lymphatic vascular growth factor therapy in combination with lymph node transfer. We have compared the therapeutic effects of different vascular endothelial growth factors (VEGF-C, VEGF-D, VEGF-C156S, and VEGF-A), in combination with lymph node transfer in mouse axilla. The lymphangiogenic effects of the growth factor therapy were examined at 3 mo postoperatively. VEGF therapy with VEGF-C and VEGF-D induced growth of new lymphatic vessels in the defect area, and VEGF-C also improved lymphatic vessel function compared with that of controls. VEGF-C156S induced moderate lymphangiogenesis, but the effect remained statistically nonsignificant. Prolymphangiogenic growth factors (VEGF-C, -D, and -C156S) also improved lymph node survival as compared with those of the VEGF-A and control group. VEGF-C, which activates both vascular endothelial growth factor receptor 2 and vascular endothelial growth factor receptor 3, gave the best therapeutic effect in this experimental lymphedema model. These results support our goal to treat secondary lymphedema by combining lymph node transfer with the growth factor therapy. VEGF-C provides the preferred alternative for growth factor therapy of lymphedema when compared with other VEGF-family growth factors, due to the superior lymphangiogenic response and minor blood vascular effects.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Xylazine hydrochloride, ≥99.0% (HPLC)
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid