Merck
Anatomical organization of the rat organum vasculosum laminae terminalis.
American journal of physiology. Regulatory, integrative and comparative physiology (2015-05-29)
Masha Prager-Khoutorsky, Charles W Bourque
ABSTRAKT
The organum vasculosum of the laminae terminalis (OVLT) is a circumventricular organ located along the ventral part of the anterior wall of the third ventricle. Because it lacks a complete blood-brain barrier (BBB), blood-borne signals detected in the OVLT provide the brain with information from the periphery and contribute to the generation of centrally mediated responses to humoral feedback and physiological stressors. Experimental studies on the rat OVLT are hindered by a poor understanding of its precise anatomical dimensions and cellular organization. In this study, we use histological techniques to characterize the spatial outline of the rat OVLT and to examine the location of neurons, astrocytes, tanycytes, and ependymocytes within its confines. Our data reveal that OVLT neurons are embedded in a dense network of tanycyte processes. Immunostaining against the neuronal marker NeuN revealed that neurons are distributed throughout the OVLT, except for a thick midline septum, which comprises densely packed cells of unknown function or lineage. Moreover, the most ventral aspect of the OVLT is devoid of neurons and is occupied by a dense network of glial cell processes that form a thick layer between the neurons and the pial surface on the ventral aspect of the nucleus. Lastly, combined detection of NeuN and c-Fos protein following systemic injection of hypertonic NaCl revealed that neurons responsive to this stimulus are located along the entire midline core of the OVLT, extending from its most anterior ventral aspect to the more caudally located "dorsal cap" region.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-NeuN Antibody, from chicken, purified by affinity chromatography