- Azide-rich peptides via an on-resin diazotransfer reaction.
Azide-rich peptides via an on-resin diazotransfer reaction.
Biopolymers (2015-03-11)
Jeannette E Marine, Xiaoli Liang, Shuang Song, Jonathan G Rudick
PMID25753459
ABSTRAKT
Azide-containing amino acids are valuable building blocks in peptide chemistry, because azides are robust partners in several bioorthogonal reactions. Replacing polar amino acids with apolar, azide-containing amino acids in solid-phase peptide synthesis can be tricky, especially when multiple azide residues are to be introduced in the amino acid sequence. We present a strategy for effectively incorporating multiple azide-containing residues site-specifically.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture