Merck
Arjunolic acid protects against DNCB-induced atopic dermatitis-like symptoms in mice by restoring a normal cytokine balance.
European cytokine network (2015-11-11)
Abdullah Alyoussef
ABSTRAKT
Atopic dermatitis (AD) is a chronically relapsing, pruritic, eczematous skin disorder accompanying allergic inflammation. AD is triggered by oxidative stress and immune imbalance. The effect of oral arjunolic acid (AA) on 2,4-dinitrochlorobenzene (DNCB)-induced atopic dermatitis in mice was investigated. Repeated epicutaneous application of DNCB to the ear and shaved dorsal skin of mice was performed to induce AD-like symptoms and skin lesions: 250mg/kg AA was given orally for three weeks to assess its anti-pruritic effects. Serum levels of tumor necrosis factor (TNF)-α, interleukin (IL)-4, IL-6, IL-10, immunoglobulin (Ig)E and caspase-3 were assessed by ELISA. We found that AA alleviated DNCB-induced AD-like symptoms as quantified by skin lesions, dermatitis score, ear thickness and scratching behavior. Levels of reactive oxygen species in the AA group were significantly inhibited compared with those in the DNCB group. In parallel, AA blocked a DNCB-induced reduction in serum levels of IL-4 and IL-10 associated with an attenuation of DNCB-induced increases in serum TNF-α, IL-6, IgE and caspase-3. The results indicate that AA suppresses DNCB-induced AD in mice via redox balance and immune modulation, and could be a safe clinical treatment for AD.