Merck
Individuals with Primary Sclerosing Cholangitis Have Elevated Levels of Biomarkers for Apoptosis but Not Necrosis.
Digestive diseases and sciences (2015-07-22)
Howard C Masuoka, Raj Vuppalanchi, Ross Deppe, Phelan Bybee, Megan Comerford, Suthat Liangpunsakul, Marwan Ghabril, Naga Chalasani
ABSTRAKT
Hepatocyte apoptosis or necrosis from accumulation of bile salts may play an important role in the disease progression of primary sclerosing cholangitis (PSC). The aim of the current study was to measure serum markers of hepatocyte apoptosis (cytokeratin-18 fragments--K18) and necrosis (high-mobility group protein B1--HMGB1) in adults with PSC and examine the relationship with disease severity. We measured serum levels of K18 and HMGB1 in well-phenotyped PSC (N = 37) and 39 control subjects (N = 39). Severity of PSC was assessed biochemically, histologically, and PSC Mayo risk score. Quantification of hepatocyte apoptosis was performed using TUNEL assay. The mean age of the study cohort was 49.7 ± 13.3 years and comprised of 67% men and 93% Caucasian. Serum K18 levels were significantly higher in the PSC patients compared to control (217.4 ± 78.1 vs. 157.0 ± 58.2 U/L, p = 0.001). However, HMGB1 levels were not different between the two groups (5.38 ± 2.99 vs. 6.28 ± 2.85 ng/mL, p = 0.15). Within the PSC group, K18 levels significantly correlated with AST (r = 0.5, p = 0.002), alkaline phosphatase (r = 0.5, p = 0.001), total bilirubin (r = 0.61, p ≤ 0.001), and albumin (r = -0.4, p = 0.02). Serum K18 levels also correlated with the level of apoptosis present on the liver biopsy (r = 0.8, p ≤ 0.001) and Mayo risk score (r = 0.4, p = 0.015). Serum K18 but not HMGB1 levels were increased in PSC and associated with severity of underlying liver disease and the degree of hepatocyte apoptosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid