Merck
  • Hydrolyzed fish proteins reduced activation of caspase-3 in H2O2 induced oxidative stressed liver cells isolated from Atlantic salmon (Salmo salar).
Hydrolyzed fish proteins reduced activation of caspase-3 in H2O2 induced oxidative stressed liver cells isolated from Atlantic salmon (Salmo salar).
SpringerPlus (2015-11-07)
M Espe, E Holen, J He, F Provan, L Chen, K B Øysæd, J Seliussen
ABSTRAKT
Hydrolyzed fish proteins (H-pro) contains high concentrations of free amino acids and low molecular peptides that potentially benefit health. The following study aimed to test whether the water soluble phase of H-pro could reduce apoptosis and inflammation in primary liver cells isolated from Atlantic salmon following H2O2 provoked oxidative stress. Cells were grown as monocultures or co-cultured with head kidney cells to assess possible cross talk in inflammation and metabolism during treatments. Cells were grown in media with or without H-pro for 2 days before being stressed with 200 µM H2O2 then harvested 24 h post exposure. Both treatments were compared to the respective treatments without H2O2 supplementation. Oxidative stressed cells had increased activation of caspase-3, but supplementation with H-pro in the media prior to the oxidative stress reduced caspase-3 activation. In conclusion, free amino acids and low molecular weight peptides from H-pro attenuated oxidative stress, and made cells able to withstand apoptosis after H2O2 provoked oxidative stress.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, FITC conjugate, 2 mg/mL, Chemicon®
Sigma-Aldrich
CelLytic MT Cell Lysis Reagent, For mammalian tissues