Merck
  • Tetramethylpyrazine reduces inflammation in liver fibrosis and inhibits inflammatory cytokine expression in hepatic stellate cells by modulating NLRP3 inflammasome pathway.
Tetramethylpyrazine reduces inflammation in liver fibrosis and inhibits inflammatory cytokine expression in hepatic stellate cells by modulating NLRP3 inflammasome pathway.
IUBMB life (2015-04-08)
Xiafei Wu, Feng Zhang, Xin Xiong, Chunfeng Lu, Naqi Lian, Yin Lu, Shizhong Zheng
ABSTRAKT
Hepatic fibrosis is concomitant with liver inflammation, which has been highlighted as significant treatment of chronic liver disease. We previously demonstrated that tetramethylpyrazine (TMP), the effective component of Ligusticum chuanxiong Hort, can inhibit the activation of HSCs and consequential anti-hepatic fibrosis. In this study, our work demonstrated that TMP improved liver histological architecture, decreased hepatic enzyme levels and attenuated collagen deposition in the rat fibrotic liver. In addition, TMP significantly protected the liver from CCl4-caused injury and fibrogenesis by suppressing inflammation with reducing levels of inflammatory cytokines, including tumor necrosis factor-α (TNF-α), NLRP3, nuclear factor-kappa B (NF-κB) and interleukin-1β (IL-1β). Experiments in vitro showed that TMP inhibited inflammatory cytokine expression in HSCs associated with disrupting platelet-derived growth factor-b receptor (PDGF-βR)/NLRP3/caspase1 pathway. These data collectively indicate that TMP can attenuate liver inflammation in liver fibrosis and possibly by targeting HSCs via PDGF-βR/NLRP3/caspase1 pathway. It provides novel mechanistic insights into TMP as a potential therapeutic remedy for hepatic fibrosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Glyburide, meets USP testing specifications
Sigma-Aldrich
Colchicine, suitable for plant cell culture, BioReagent, ≥95% (HPLC)
Sigma-Aldrich
Glybenclamide, ≥99% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)