Merck
Design of hydrophilic metal organic framework water adsorbents for heat reallocation.
Advanced materials (Deerfield Beach, Fla.) (2015-07-21)
Amandine Cadiau, Ji Sun Lee, Daiane Damasceno Borges, Paul Fabry, Thomas Devic, Michael T Wharmby, Charlotte Martineau, Damien Foucher, Francis Taulelle, Chul-Ho Jun, Young Kyu Hwang, Norbert Stock, Martijn F De Lange, Freek Kapteijn, Jorge Gascon, Guillaume Maurin, Jong-San Chang, Christian Serre
ABSTRAKT
A new hydrothermally stable Al polycarboxylate metal-organic framework (MOF) based on a heteroatom bio-derived aromatic spacer is designed through a template-free green synthesis process. It appears that in some test conditions this MOF outperforms the heat reallocation performances of commercial SAPO-34.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Allyl methyl sulfone, 96%