Merck
Ultrasmall Chitosan-Genipin Nanocarriers Fabricated from Reverse Microemulsion Process for Tumor Photothermal Therapy in Mice.
Biomacromolecules (2015-06-16)
Xiaojie Song, Hao Wu, Shen Li, Yanfang Wang, Xiaojun Ma, Mingqian Tan
ABSTRAKT
Nanocarriers play an important role in improving the photo- and thermal-stability of photosensitizers to gain better pharmacokinetics behavior in tumor photothermal therapy. Herein, PEGylated chitosan (CG-PEG; PEG: polyethylene glycol) nanoparticles with ultrasmall size (∼5 nm) were prepared through a water-in-oil reverse microemulsion method using genipin as a cross-linker. Particle size and zeta-potential can be tuned by varying the molar ratio between chitosan amino groups and genipin. CG-PEG-ICG (ICG: indocyanine green) nanoparticles were fabricated by adding ICG to CG-PEG aqueous solution through a self-assembly method via electrostatic interaction. The resultant CG-PEG-ICG nanoparticles exhibited improved photo- and thermal-stability, good biocompatibility, and low toxicity. When irradiated with a laser, the cells incubated with CG-PEG-ICG nanoparticles showed very low cell viability (15%), indicating the CG-PEG-ICG nanoparticles possess high in vitro photothermal toxicity. Moreover, the CG-PEG nanocarriers can significantly alter the biodistribution and prolong the retention time of ICG in the mice body after intravenous injection. In vivo photothermal study of tumors injected with CG-PEG-ICG nanoparticles containing ICG at a concentration greater than 100 μg·mL(-1) (100 μL) induced irreversible tissue damage. The growth of U87 tumors was dramatically inhibited by CG-PEG-ICG nanoparticles, demonstrating that the CG-PEG nanoparticles may act as potential ICG nanocarriers for effective in vivo tumor photothermal therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Triton X-100, laboratory grade