Merck
High Amylose Starch with Low In Vitro Digestibility Stimulates Hindgut Fermentation and Has a Bifidogenic Effect in Weaned Pigs.
The Journal of nutrition (2015-09-18)
Janelle M Fouhse, Michael G Gänzle, Prajwal R Regmi, Theo A T G van Kempen, Ruurd T Zijlstra
ABSTRAKT
Dietary amylose resists enzymatic digestion, thereby providing a substrate for microbial fermentation that stimulates proliferation of beneficial microbiota and production of short-chain fatty acids (SCFAs) in the large intestine of pigs and humans. However, the effect of increasing dietary amylose in pigs immediately postweaning on growth, nutrient digestibility and flow, and intestinal microbial and SCFA profiles has not been studied and can be used as a model for newly weaned human infants. We studied the effects of increasing dietary amylose on growth, nutrient digestibility, and intestinal microbial and metabolite profiles in weaned pigs. Weaned pigs (n = 32) were randomly allocated to 1 of 4 diets containing 67% starch with 0%, 20%, 28%, or 63% amylose for 21 d. Subsequently, pigs were killed to collect feces and digesta for measuring starch digestion and microbial and metabolite profiles. Feeding weaned pigs 63% compared with 0%, 20%, and 28% amylose decreased (P < 0.05) feed intake by 5% and growth by ≥ 12%. Ileal digestibility of dry matter decreased (P < 0.05) by 10% and starch by 9%, thereby increasing (P < 0.05) hindgut fermentation, cecal and colonic total SCFAs, and colonic Bacteroides, and lowering (P < 0.01) ileal, cecal, and colonic pH in pigs consuming 63% compared with 0%, 20%, and 28% amylose. Cecal and colonic Bifidobacteria spp. increased by 14-30% (P < 0.05) and Clostridium clusters IV and XIVa were decreased (P < 0.01) in pigs consuming 63% compared with 0%, 20%, and 28% amylose. Increasing dietary amylose in pigs immediately postweaning stimulated hindgut fermentation and Bifidobacteria spp., thereby manipulating the gut environment, but also reduced intake and growth. An optimum dietary amylose concentration should be determined, which would maintain desired growth rate and gut environment in weaned pigs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis