Merck
On the pH dependence of class-1 RF-dependent termination of mRNA translation.
Journal of molecular biology (2015-01-27)
Gabriele Indrisiunaite, Michael Y Pavlov, Valérie Heurgué-Hamard, Måns Ehrenberg
ABSTRAKT
We have studied the pH dependence of the rate of termination of bacterial protein synthesis catalyzed by a class-1 release factor (RF1 or RF2). We used a classical quench-flow technique and a newly developed stopped-flow technique that relies on the use of fluorescently labeled peptides. We found the termination rate to increase with increasing pH and, eventually, to saturate at about 70 s(-1) with an apparent pKa value of about 7.6. From our data, we suggest that class-1 RF termination is rate limited by the chemistry of ester bond hydrolysis at low pH and by a stop-codon-dependent and pH-independent conformational change of RFs at high pH. We propose that RF-dependent termination depends on the participation of a hydroxide ion rather than a water molecule in the hydrolysis of the ester bond between the P-site tRNA and its peptide chain. We provide a simple explanation for why the rate of termination saturated at high pH in our experiments but not in those of others.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Spermidine, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phospho(enol)pyruvic acid trisodium salt hydrate, ≥97% (enzymatic)
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Spermidine, BioReagent, Molecular Biology, suitable for cell culture, ≥98%
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis
Sigma-Aldrich
1,4-Diaminobutane, 99%