Merck
  • Discovery of a Novel Series of Imidazo[1,2-a]pyrimidine Derivatives as Potent and Orally Bioavailable Lipoprotein-Associated Phospholipase A2 Inhibitors.
Discovery of a Novel Series of Imidazo[1,2-a]pyrimidine Derivatives as Potent and Orally Bioavailable Lipoprotein-Associated Phospholipase A2 Inhibitors.
Journal of medicinal chemistry (2015-10-20)
Xinde Chen, Wenwei Xu, Kai Wang, Mingguang Mo, Wei Zhang, Lili Du, Xiaojing Yuan, Yechun Xu, Yiping Wang, Jianhua Shen
ABSTRAKT
Inhibition of lipoprotein-associated phospholipase A2 (Lp-PLA2) has been suggested to be a promising therapeutic strategy for several inflammation-associated diseases, including atherosclerosis, Alzheimer's disease, and diabetic macular edema. Herein, we report the discovery of a novel series of Lp-PLA2 inhibitors constructed on an imidazo[1,2-a]pyrimidine scaffold through a conformational restriction strategy. Structure-activity relationship (SAR) analysis resulted in the identification of several compounds with high potency in vitro and good metabolic stability in liver S9 fractions. Compounds 7c and 14b selected for further exploration in vivo demonstrated excellent pharmacokinetic profiles and exhibited significant inhibitory efficacy in SD rats upon oral dosing.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Trioctylphosphine, technical grade, 90%
Sigma-Aldrich
Trioctylphosphine, 97%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O