Merck
Bioinspired production of magnetic laccase-biotitania particles for the removal of endocrine disrupting chemicals.
Biotechnology and bioengineering (2015-06-11)
Inés Ardao, Delphine Magnin, Spiros N Agathos
ABSTRAKT
Microbial laccases are powerful enzymes capable of degrading lignin and other recalcitrant compounds including endocrine disrupting chemicals (EDCs). Efficient EDC removal on an industrial scale requires robust, stable, easy to handle and cost-effective immobilized biocatalysts. In this direction, magnetic biocatalysts are attractive due to their easy separation through an external magnetic field. Recently, a bioinspired immobilization technique that mimics the natural biomineralization reactions in diatoms has emerged as a fast and versatile tool for generating robust, cheap, and highly stable (nano) biocatalysts. In this work, bioinspired formation of a biotitania matrix is triggered on the surface of magnetic particles in the presence of laccase in order to produce laccase-biotitania (lac-bioTiO2 ) biocatalysts suitable for environmental applications using a novel, fast and versatile enzyme entrapment technique. Highly active lac-bioTiO2 particles have been produced and the effect of different parameters (enzyme loading, titania precursor concentration, pH, duration of the biotitania formation, and laccase adsorption steps) on the apparent activity yield of these biocatalysts were evaluated, the concentration of the titania precursor being the most influential. The lac-bioTiO2 particles were able to catalyze the removal of bisphenol A, 17α-ethinylestradiol and diclofenac in a mixture of six model EDCs and retained 90% of activity after five reaction cycles and 60% after 10 cycles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%