Merck
  • Brominated polyunsaturated lipids with protein tyrosine phosphatase-1B inhibitory activity from Chinese marine sponge Xestospongia testudinaria.
Brominated polyunsaturated lipids with protein tyrosine phosphatase-1B inhibitory activity from Chinese marine sponge Xestospongia testudinaria.
Journal of Asian natural products research (2015-04-03)
Wen-Fei He, Lin-Fu Liang, You-Sheng Cai, Li-Xin Gao, Yu-Fen Li, Jia Li, Hai-Li Liu, Yue-Wei Guo
ABSTRAKT
A new brominated polyunsaturated lipid, methyl (E,E)-14,14-dibromo-4,6,13-tetradecatrienoate (1), along with three known related analogues (2-4), were isolated from the Et2O-soluble portion of the acetone extract of Chinese marine sponge Xestospongia testudinaria treated with diazomethane. The structure of the new compound was elucidated by detailed spectroscopic analysis and by comparison with literature data. Compound 3 exhibited significant inhibitory activity against protein tyrosine phosphatase 1B (PTP1B), a key target for the treatment of type II diabetes and obesity, with an IC50 value of 5.30 ± 0.61 μM, when compared to the positive control oleanolic acid (IC50 = 2.39 ± 0.26 μM).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%