Merck
  • Dietary peppermint (Mentha piperita) extracts promote growth performance and increase the main humoral immune parameters (both at mucosal and systemic level) of Caspian brown trout (Salmo trutta caspius Kessler, 1877).
Dietary peppermint (Mentha piperita) extracts promote growth performance and increase the main humoral immune parameters (both at mucosal and systemic level) of Caspian brown trout (Salmo trutta caspius Kessler, 1877).
Fish & shellfish immunology (2015-10-13)
Milad Adel, Reza Safari, Reza Pourgholam, Jalil Zorriehzahra, Maria Ángeles Esteban
ABSTRAKT
The effects of dietary administration of peppermint (Mentha piperita L.) on Caspian brown trout fish (Salmo trutta caspius) were studied. Fish were divided into 4 groups before being fed diets supplemented with 0% (control), 1%, 2% and 3% of peppermint extracts for 8 weeks. Dose-dependent increases in growth, immune (both in skin mucus and blood serum) and hematological parameters (number of white cells, hematocrit and hemoglobin content), as well as in amylase activity and in the number of lactic acid bacteria on intestine were recorded in fish fed supplemented diets compared to control fish. However, the dietary peppermint supplements have different effects on the number of blood leucocytes depending on the leukocyte cell type. While no significant differences were observed in the number of blood monocytes and eosinophils, the number of lymphocytes was decreased, respectively, on fish fed peppermint enriched diets, respect to the values found in control fish. Furthermore, dietary peppermint supplements have no significant effect on blood biochemical parameters, enzymatic activities of liver determined in serum and total viable aerobic bacterial count on intestine of Caspian brown trout. Present results support that dietary administration of peppermint promotes growth performance and increases the main humoral immune parameters (both at mucosal and systemic level) and the number of the endogenous lactic acid bacteria of Caspian brown trout. This study underlying several positive effects of dietary administration of peppermint to farmed fish.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Sodium phosphate, 96%