Merck
A Study of the 3,3,3-Trinitropropyl Unit as a Potential Energetic Building Block.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-09-18)
Quirin J Axthammer, Burkhard Krumm, Thomas M Klapötke, Regina Scharf
ABSTRAKT
Compared with the well-established 2,2,2-trinitroethyl group in the chemistry of energetic materials, the 3,3,3-trinitropropyl group is less investigated regarding its chemical and energetic properties. Thus, investigations on the syntheses of several compounds containing the 3,3,3-trinitropropyl group were performed and their properties compared with the 2,2,2-trinitroethyl group. All materials were thoroughly characterized, including single-crystal X-ray diffraction studies. The thermal stabilities were examined using differential thermal analysis (DSC) and the sensitivities towards impact, friction, and electrostatic discharge were tested using a drop hammer, a friction tester, and an electrical discharge device. The energies of formation were calculated and several detonation parameters such as the velocity of detonation and the propulsion performance were estimated with the program package EXPLO5.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor