Merck
New tools for chloroplast genetic engineering.
Nature biotechnology (1999-09-03)
H Daniell
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Spectinomycin, Ready Made Solution, 100 mg/mL in DMSO/H2O, 1:1