Merck
In vitro antioxidant profiling of seabuckthorn varieties and their adaptogenic response to high altitude-induced stress.
International journal of biometeorology (2014-11-12)
Priyanka Sharma, Geetha Suryakumar, Virendra Singh, Kshipra Misra, Shashi Bala Singh
ABSTRAKT
In the past few years, seabuckthorn plants have gained special attention due to their ability to grow in the harshest of the environment. This adaptability may be contributed by various antioxidants present in the plants besides other morphological adaptation. As in vivo studies cannot be justified without in vitro studies, the present investigation carried out evaluation of both in vitro and in vivo antioxidant potentials of aqueous and alcoholic extracts of the leaves of Hippophae salicifolia (HS) and Hippophae rhamnoides mongolica (HRM) in comparison with Hippophae rhamnoides turkestanica (HRT). The results had clearly depicted that in vitro antioxidant potential of the extracts was responsible for the in vivo adaptogenic performance in animals during cold and hypoxia exposure under restraint stress. Total phenolic content (TPC), total flavonoid content (TFC), total protein content, and antioxidant potential were determined. For adaptogenic studies, rats with oral drug supplementation were exposed to Cold-hypoxia-restraint (C-H-R) stresses-induced hypothermia, as a measure of endurance. Aqueous extracts of HS showed maximum (99 %) resistance compared to HRT (81 %) and HRM (29 %). The levels of biochemical parameters such as malondialdehyde (MDA), reactive oxygen species (ROS), lactate dehydrogenase (LDH), superoxide dismutase (SOD), glutathione (GSH/GSSG), and catalase (CAT) in blood samples also revealed that the aqueous leaf extract of HS has better antioxidant and adaptogenic potential compared to HRM.