Merck
Reproductive Stage and Modulation of Stress-Induced Tau Phosphorylation in Female Rats.
Journal of neuroendocrinology (2015-10-29)
Danielle Steinmetz, Eugenia Ramos, Shannon N Campbell, Teresa Morales, Robert A Rissman
ABSTRAKT
Chronic stress is implicated as a risk factor for Alzheimer's disease (AD) and other neurodegenerative disorders. Although the specific mechanisms linking stress exposure and AD vulnerability have yet to be fully determined, our laboratory and others have shown that acute and repeated restraint stress in rodents leads to an increase in hippocampal tau phosphorylation (tau-P) and tau insolubility, a critical component of tau pathology in AD. Although tau phosphorylation induced by acute psychological stress is dependent on intact signaling through the type 1 corticotropin-releasing factor receptor, how sex steroids or other modulators contribute to this effect is unknown. A naturally occurring attenuation of the stress response is observed in female rats at the end of pregnancy and throughout lactation. To test the hypothesis that decreased sensitivity to stress during lactation modulates stress-induced tau-P, cohorts of virgin, lactating and weaned female rats were subjected to 30 min of restraint stress or no stress (control) and were killed 20 min or 24 h after the episode. Exposure to restraint stress induced a significant decrease in tau-P in the hippocampus of lactating rats killed 20 min after stress compared to lactating controls and virgins subjected to stress treatment. Lactating rats killed 24 hr after restraint stress exposure showed significant elevation in tau-P compared to lactating cohorts killed 20 min after stress. Levels of tau-P in these latter cohorts did not differ signficantly from control animals. Furthermore, glycogen synthase kinase (GSK)3-α levels were significantly decreased in stressed lactating animals at both timepoints. This suggests a steep, yet transient stress-induced dephosphorylation of tau, influenced by GSK3, in the hippocampus of lactating rats.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Corticosterone, ≥92%