Merck
Non-classical azoreductase secretion in Clostridium perfringens in response to sulfonated azo dye exposure.
Anaerobe (2015-04-18)
Jessica M Morrison, Gilbert H John
ABSTRAKT
Clostridium perfringens, a strictly anaerobic microorganism and inhabitant of the human intestine, has been shown to produce an azoreductase enzyme (AzoC), an NADH-dependent flavin oxidoreductase. This enzyme reduces azo dyes into aromatic amines, which can be carcinogenic. A significant amount of work has been completed on the activity of AzoC. Despite this, much is still unknown, including whether azoreduction of these dyes occurs intracellularly or extracellulary. A physiological study of C. perfringens involving the effect of azo dye exposure was completed to answer this question. Through exposure studies, azo dyes were found to cause cytoplasmic protein release, including AzoC, from C. perfringens in dividing and non-dividing cells. Sulfonation (negative charge) of azo dyes proved to be the key to facilitating protein release of AzoC and was found to be azo-dye-concentration-dependent. Additionally, AzoC was found to localize to the Gram-positive periplasmic region. Using a ΔazoC knockout mutant, the presence of additional azoreductases in C. perfringens was suggested. These results support the notion that the azoreduction of these dyes may occur extracellularly for the commensal C. perfringens in the intestine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium benzenesulfonate, 97%
Sigma-Aldrich
Methyl Red, ACS reagent, crystalline
Sigma-Aldrich
Sodium phosphate, 96%