Merck
Accurate assessment of the biodegradation of cationic surfactants in activated sludge reactors (OECD TG 303A).
Ecotoxicology and environmental safety (2015-04-29)
Roy Geerts, Cornelis G van Ginkel, Caroline M Plugge
ABSTRAKT
The continuous-fed activated sludge test (OECD TG 303A) was used to predict the removal of cationic surfactants from wastewater in activated sludge plants. However, a method to differentiate between adsorption and biodegradation is not provided in these guidelines. Assessment of removal by biodegradation was possible with analysis of the surfactant present in mixed liquid suspended solids in combination with a simple equation. This equation was derived from the mass balance of the activated sludge unit in steady state. The removal by biodegradation of decylamine, tetradecylamine, octadecylamine, dioctadecylmethylamine and dioctadecyldimethylammonium chloride that have different capacities to adsorb was >99.9%, >99.9%, 98.2%, 94.2%, and 69.0%, respectively. The total removal of all five cationic surfactants from the influent was ≥98.8%. The removal of octadecylamine spiked at different influent concentrations indicated first order kinetics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Decylamine, 95%
Sigma-Aldrich
Decylamine, ≥99.0% (GC)
Sigma-Aldrich
Octadecylamine, ≥99.0% (GC)
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Sigma-Aldrich
Tetradecylamine, 95%
Sigma-Aldrich
Octadecylamine, technical grade, 90%