Merck
Influence of in ovo mercury exposure, lake acidity, and other factors on common loon egg and chick quality in Wisconsin.
Environmental toxicology and chemistry (2015-06-23)
Kevin P Kenow, Michael W Meyer, Ronald Rossmann, Brian R Gray, Michael T Arts
ABSTRAKT
A field study was conducted in Wisconsin (USA) to characterize in ovo mercury (Hg) exposure in common loons (Gavia immer). Total Hg mass fractions ranged from 0.17 µg/g to 1.23 µg/g wet weight in eggs collected from nests on lakes representing a wide range of pH (5.0-8.1) and were modeled as a function of maternal loon Hg exposure and egg laying order. Blood total Hg mass fractions in a sample of loon chicks ranged from 0.84 µg/g to 3.86 µg/g wet weight at hatch. Factors other than mercury exposure that may have persistent consequences on development of chicks from eggs collected on low-pH lakes (i.e., egg selenium, calcium, and fatty acid mass fractions) do not seem to be contributing to reported differences in loon chick quality as a function of lake pH. However, it was observed that adult male loons holding territories on neutral-pH lakes were larger on average than those occupying territories on low-pH lakes. Differences in adult body size of common loons holding territories on neutral-versus low-pH lakes may have genetic implications for differences in lake-source-related quality (i.e., size) in chicks. The tendency for high in ovo Hg exposure and smaller adult male size to co-occur in low-pH lakes complicates the interpretation of the relative contributions of each to resulting chick quality.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tin(II) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(II) chloride, ≥99.99% trace metals basis