Merck
  • Green approach to stereoselective synthesis of benzo[d]chromeno[3,4- h]oxathiazonine derivatives via MCRs in water: a combined experimental and DFT study.
Green approach to stereoselective synthesis of benzo[d]chromeno[3,4- h]oxathiazonine derivatives via MCRs in water: a combined experimental and DFT study.
Combinatorial chemistry & high throughput screening (2015-09-16)
Hoorieh Djahaniani, Farzaneh Fatemi, Fatemeh Ektefa, Bita Mohtat, Behrouz Notash
ABSTRAKT
(7S, 14S, 16R)- dialkyl 6-oxo-6,7,13,14-tetrahydro-7,14-methanobenzo[d]chromeno[3,4- h][1,6,3]oxathiazo-nine-14,16-dicarboxylates 4 and (S)-methyl 2-(2-methoxy-2-oxoethyl)-2,3- dihydrobenzo[d]thiazole-2-carboxylates 5 were readily synthesized in a ratio 3:1 and moderated yield by multicomponent reactions of 4-hydroxycoumarin and acetylenic esters with benzothiazole without using a catalyst. Also, The GIAO/DFT approach at the B3LYP/6-31G** level of theory was used to calculate the (1)H, (15)N, (17)O and (13)C NMR chemical shifts of product 4b. These computations are performed on the basis of X-ray structural data which are collected at 120(2) K temperature. In order to take into account intermolecular hydrogen bonds and the van der Waals effects, two different sizes of clusters (two model of trimeric and pentameric clusters) have been considered. A comparison between the experimental (Exp.) and calculated (Cal.) (1)H and (13)C NMR chemical shifts may reveal that the solution contains monomer, trimer1, trimer2, and pentamer models.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG