Merck
  • The impact of two different inspiratory to expiratory ratios (1:1 and 1:2) on respiratory mechanics and oxygenation during volume-controlled ventilation in robot-assisted laparoscopic radical prostatectomy: a randomized controlled trial.
The impact of two different inspiratory to expiratory ratios (1:1 and 1:2) on respiratory mechanics and oxygenation during volume-controlled ventilation in robot-assisted laparoscopic radical prostatectomy: a randomized controlled trial.
Canadian journal of anaesthesia = Journal canadien d'anesthesie (2015-04-15)
Min-Soo Kim, Na Young Kim, Ki-Young Lee, Young Deuk Choi, Jung Hwa Hong, Sun-Joon Bai
ABSTRAKT
Volume-controlled ventilation with a prolonged inspiratory to expiratory ratio (I:E ratio) has been used to optimize gas exchange and respiratory mechanics in various surgical settings. We hypothesized that, when compared with an I:E ratio of 1:2, a prolonged I:E ratio of 1:1 would improve respiratory mechanics without reducing cardiac output (CO) during pneumoperitoneum and steep Trendelenburg positioning, both of which can impair respiratory function in robot-assisted laparoscopic radical prostatectomy. Furthermore, we evaluated its effect on oxygenation during robot-assisted laparoscopic radical prostatectomy. Eighty patients undergoing robot-assisted laparoscopic radical prostatectomy were randomly allocated to receive an I:E ratio of either 1:1 (group 1:1) or 1:2 (group 1:2). The primary endpoint, peak airway pressure (Ppeak), as well as hemodynamic data, including cardiac output (CO) and arterial oxygen tension (PaO2), were compared between groups at four time points: ten minutes after anesthesia induction (T1), 30 and 60 min after pneumoperitoneum with steep Trendelenburg positioning (T2 and T3), and ten minutes after supine positioning (T4). Overall comparisons were made between groups using linear mixed model analysis with post hoc testing of individual time points adjusted using a Bonferroni correction. Linear mixed model analysis showed a significant overall difference in Ppeak between the two groups (P < 0.001). Post hoc analysis showed a significantly lower mean (SD) Ppeak in group 1:1 than in group 1:2 at T2 [28.4 (4.0) cm H2O vs 32.8 (5.2) cm H2O, respectively; mean difference, 4.3 cm H2O; 95% confidence interval (CI), 2.3 to 6.4; P < 0.001] and T3 [27.8 (3.9) cm H2O vs 32.6 (5.0) cm H2O, respectively; mean difference, 4.7 cm H2O; 95% CI, 2.7 to 6.7; P < 0.001]. The CO assessed over these time points was comparable in both groups (P = 0.784). In addition, there were no significant differences in PaO2 between the two groups (P = 0.521). Compared with an I:E ratio of 1:2, a ratio of 1:1 lowered Ppeak without reducing CO during pneumoperitoneum and steep Trendelenburg positioning. Nevertheless, our results did not support its use solely for improving oxygenation. This trial was registered at http://clinicaltrials.gov/ (NCT01892449).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Molecular Biology, sterile filtered