Merck
  • Role of digital tomosynthesis and dual energy subtraction digital radiography in detection of parenchymal lesions in active pulmonary tuberculosis.
Role of digital tomosynthesis and dual energy subtraction digital radiography in detection of parenchymal lesions in active pulmonary tuberculosis.
European journal of radiology (2015-06-14)
Madhurima Sharma, Manavjit Singh Sandhu, Ujjwal Gorsi, Dheeraj Gupta, Niranjan Khandelwal
ABSTRAKT
To assess the role of digital tomosynthesis (DTS) and dual energy subtraction digital radiography (DES-DR) in detection of parenchymal lesions in active pulmonary tuberculosis (TB) and to compare them with digital radiography (DR). This prospective study was approved by our institutional review committee. DTS and DES-DR were performed in 62 patients with active pulmonary TB within one week of multidetector computed tomography (MDCT) study. Findings of active pulmonary TB, that is consolidation, cavitation and nodules were noted on digital radiography (DR), DTS and DES-DR in all patients. Sensitivity, specificity, positive and negative predictive values of all 3 modalities was calculated with MDCT as reference standard. In addition presence of centrilobular nodules was also noted on DTS. Our study comprised of 62 patients (33 males, 29 females with age range 18-82 years). Sensitivity and specificity of DTS for detection of nodules and cavitation was better than DR and DES-DR. Sensitivity and specificity of DTS for detection of consolidation was comparable to DR and DES-DR. DES-DR performed better than DR in detection of nodules and cavitation. DTS was also able to detect centrilobular nodules with sensitivity and specificity of 57.4% and 86.5% respectively. DTS and DES-DR perform better than DR in detection of nodules, consolidation and cavitation in pulmonary TB. DTS gives better results than DES-DR, particularly in detection of cavitation and has moderate sensitivity for detection of centrilobular nodules. Thus DTS can be used for evaluation of patients of suspected pulmonary TB, thereby giving a more confident diagnosis of active disease and also in follow up.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cesium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cesium iodide, 99.999% trace metals basis
Sigma-Aldrich
Cesium iodide, 99.9% trace metals basis