Merck
Enantio- and Diastereoselective Formal Hetero-Diels-Alder Reactions of Trifluoromethylated Enones Catalyzed by Chiral Primary Amines.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-17)
Yong-Jun Lin, Li-Na Du, Tai-Ran Kang, Quan-Zhong Liu, Ze-Qin Chen, Long He
ABSTRAKT
Enantioselective formal hetero-Diels-Alder reactions of trifluoromethylated enones and 2-amino-1,3-butadienes generated in situ from aliphatic acyclic enones and chiral primary amines are reported. The corresponding tetrahydropyran-4-ones are formed in up to 94 % yield and with up to 94 % ee. The reaction was carried out through a stepwise mechanism, including initial aminocatalytic aldol condensation of 2-amino-1,3-butadiene to the trifluoromethylated carbonyl group followed by an intramolecular oxa-Michael addition. Both NMR investigation and theoretical calculations on the transition state indicate that the protonated tertiary amine could effectively activate the carbonyl group of the trifluoromethyl ketone to promote the addition process through hydrogen-bonding interaction of N-H⋅⋅⋅F and N-H⋅⋅⋅O simultaneously, and thus provide a chiral environment for the approach of amino-1,3-butadienes to the activated trifluoromethyl ketone, resulting in high enantioselectivity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS