Merck
Identification of ageing-associated naturally occurring peptides in human urine.
Oncotarget (2015-10-03)
Esther Nkuipou-Kenfack, Akshay Bhat, Julie Klein, Vera Jankowski, William Mullen, Antonia Vlahou, Mohammed Dakna, Thomas Koeck, Joost P Schanstra, Petra Zürbig, Karl L Rudolph, Björn Schumacher, Andreas Pich, Harald Mischak
ABSTRAKT
To assess normal and pathological peptidomic changes that may lead to an improved understanding of molecular mechanisms underlying ageing, urinarypeptidomes of 1227 healthy and 10333 diseased individuals between 20 and 86 years of age were investigated. The diseases thereby comprised diabetes mellitus, renal and cardiovascular diseases. Using age as a continuous variable, 116 peptides were identified that significantly (p < 0.05; |ρ|≥0.2) correlated with age in the healthy cohort. The same approach was applied to the diseased cohort. Upon comparison of the peptide patterns of the two cohorts 112 common age-correlated peptides were identified. These 112 peptides predominantly originated from collagen, uromodulin and fibrinogen. While most fibrillar and basement membrane collagen fragments showed a decreased age-related excretion, uromodulin, beta-2-microglobulin and fibrinogen fragments showed an increase. Peptide-based in silico protease analysis was performed and 32 proteases, including matrix metalloproteinases and cathepsins, were predicted to be involved in ageing. Identified peptides, predicted proteases and patient information were combined in a systems biology pathway analysis to identify molecular pathways associated with normal and/or pathological ageing. While perturbations in collagen homeostasis, trafficking of toll-like receptors and endosomal pathways were commonly identified, degradation of insulin-like growth factor-binding proteins was uniquely identified in pathological ageing.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
E-Toxate Water, endotoxin, free