Merck
RLN2 Is a Positive Regulator of AKT-2-Induced Gene Expression Required for Osteosarcoma Cells Invasion and Chemoresistance.
BioMed research international (2015-08-01)
Jinfeng Ma, Hai Huang, Zenggang Han, Changzheng Zhu, Bin Yue
ABSTRAKT
The aim of the study was to determine the effect of H2 relaxin (RLN2) on invasion, migration, and chemosensitivity to cisplatin in human osteosarcoma U2-OS and MG-63 cells and then to investigate the effect of RLN2 on the AKT/NF-κB signaling pathway. The expression of RLN2, p-AKT (Ser473), and p-ERK1/2 (Phospho-Thr202/Tyr204) proteins was detected by western blot in OS tissues from 21 patients with pulmonary metastatic disease, and the correlation between RLN2 and p-AKT or RLN2 and p-ERK1/2 expression was investigated. RLN2 expression was inhibited by RLN2 siRNA transfection in the MG-63 cells. RLN2 was overexpressed in the U2-OS cells by treatment with recombinant relaxin. The results showed that positive relation was found between RLN2 and p-AKT expression in tissues of OS. Silencing RLN2 inhibited cell migratory and invasive ability and angiogenesis formation and increased the chemosensitivity to cisplatin in MG-63 cells. RLN2 overexpression promoted migratory and invasive ability and angiogenesis and increased the chemoresistance to cisplatin in U2-OS cells. Silencing RLN2 inhibited the activity of AKT/NF-κB signaling pathway in MG-63 cells, and vice versa. Blockage of both pathways by specific inhibitors abrogated RLN2-induced survival and invasion of OS cells, and vice versa. Our results indicated RLN2 confers to migratory and invasive ability, angiogenesis, and chemoresistance to cisplatin via modulating the AKT/NF-κB signaling pathway in vitro.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting human RLN2
Sigma-Aldrich
N-(3-Aminopropyl)methacrylamide hydrochloride, contains ≤1,000 ppm MEHQ as stabilizer, 98% (HPLC)
Sigma-Aldrich
4-Aminophenylmercuric acetate, ≥90% (titration)
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline