Merck
Structural and magnetic properties of mixed Co-Ln (Ln = Nd, Sm, Eu, Gd and Ho) diethyleneglycolate complexes.
Dalton transactions (Cambridge, England : 2003) (2015-08-20)
A Abdallah, T Gaudisson, R Sibille, S Nowak, W Cheikhrouhou-Koubaa, K Shinoda, M François, S Ammar
ABSTRAKT
New hybrid compounds LnCoCl(deg)2 (deg = diethyleneglycolate; Ln = Nd, Sm, Eu, Gd and Ho) have been synthesized by mixing cobalt and rare earth cations in a boiling diethyleneglycol (degH2) medium. Their crystallographic structures have been ab initio solved from synchrotron powder diffraction data. They consist of edge sharing tetrameric sub-units [(Ln2Co2)(deg)4(Cl)2] forming 1D infinite chains along the c parameter of a monoclinic unit cell (SG = C2/c). The five- and seven-coordination of Co(2+) and Ln(3+) cations inferred from the crystallographic results is confirmed by UV-visible absorption and Extended X-ray Absorption Fine Structure (EXAFS) spectroscopy. In the LnCoCl(deg)2 (Ln = Nd, Sm, Eu, Gd, Ho) series, weak antiferromagnetic superexchange interactions have been evidenced, between high spin Co(2+) and Ln(3+) orbitally degenerate cations. These materials are considered as potential precursors for the simultaneous reduction of Co-Ln-glycolate species into bimetallic nanoparticles by the polyol process.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%