Merck
¹³C SPE MAS measurement of ligand concentration in compressible chromatographic beads.
Magnetic resonance in chemistry : MRC (2015-06-09)
Fredrik Elwinger, Sergey V Dvinskikh, István Furó
ABSTRAKT
A method for measuring the ligand concentration in heterogeneous materials like chromatography media is described. In this method, (13)C single pulse excitation magic angle spinning NMR experiment with broadband (1)H decoupling is used to determine the peak integrals for a butyl ligand in the spectrum of a dried chromatography medium. Within a carefully controlled protocol, those integrals compared with that of the internal reference compound dimethyl sulfone provide the required volume concentration with an accuracy of ca 2%. The effects of temperature, degree of hydration, and other experimental parameters are discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfone, 98%
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Supelco
3-(Trimethylsilyl)propionic-2,2,3,3-d4 acid sodium salt, 98 atom % D
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
meso-2,3-Dimercaptosuccinic acid, ~98%
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Sigma-Aldrich
Dimethyl sulfide, ≥99%