Merck
Synthesis and receptor binding assay of indolin-2-one derivatives as dopamine D4 receptor ligands.
Die Pharmazie (2015-09-19)
Gu-cai Li, Bian-ling Zhang, Jiao-yun Xia, Zheng-Jun Fang
ABSTRAKT
Five indolin-2-one derivatives bearing piperazinylbutyl side chains attached to the amide nitrogen were synthesized from 2-indolinone. 1-(4-Bromobutyl)-indolin-2-one was reacted with 1-piperazinecarboxaldehyde to form 1-(4-(4-formyl-1-piperazinyl)butyl)indolin-2-one (2). In the presence of H2SO4, the aldehyde moiety was removed from 1-(4-(4-formyl-1-piperazinyl)butyl)indolin-2-one and then 1-(4-(1-piperazinyl)butyl)indolin-2-one (3) was obtained, this compound was reacted with benzaldehyde derivatives to give the target compounds 4 a-e by N-alkylation reaction. The structures of the intermediates and the target compounds were characterized by 1H NMR, ESI-MS spectra and elemental analyses. In vitro receptor binding assays at D2, D3, D4 receptor subtypes of the target compounds were performed and the five compounds showed selectivity towards D2-like receptors. Among them, 1-(4-(4-(4-hydroxybenzy)-1-piperazinyl)butyl) indolin-2-one (4c) exhibited a remarkable affinity and selectivity to D4 receptor with K(i) value of 0.5 nM. The results indicated that 1-(4-(4-(4-hydroxybenzy)-1-piperazinyl)butyl)indolin-2-one might be a potential dopamine D4 receptor ligand.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Oxindole, 97%
Sigma-Aldrich
1,4-Dibromobutane, 99%
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥97%, FG
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Sigma-Aldrich
4-Hydroxybenzaldehyde, 98%
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥95.0% (HPLC)
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Chlorobenzaldehyde, 97%