Merck
  • Assessment of physiological and biochemical responses, metal tolerance and accumulation in two eucalypt hybrid clones for phytoremediation of cadmium-contaminated waters.
Assessment of physiological and biochemical responses, metal tolerance and accumulation in two eucalypt hybrid clones for phytoremediation of cadmium-contaminated waters.
Journal of environmental management (2015-08-09)
Fabrizio Pietrini, Valentina Iori, Daniele Bianconi, Giovanni Mughini, Angelo Massacci, Massimo Zacchini
ABSTRAKT
Eucalyptus is a promising species for ecological restoration but plant performances under environmental constraints need to be better investigated. In particular, the toxic effects of metals on this plant species are poorly described in the literature. In this work, morpho-physiological and biochemical responses to cadmium were analysed in two eucalypt genotypes (hybrid clones of Eucalyptus camaldulensis × Eucalyptus globulus ssp. bicostata J.B. Kirkp named Velino ex 7 and Viglio ex 358) exposed for 3 weeks to 50 μM CdSO4 under hydroponics. The two eucalypt clones showed a different sensitivity to the metal. The growth reduction caused by cadmium was less than 30% in clone Velino and about 50% in clone Viglio. Cadmium mostly accumulated in plant roots and, to a lesser extent, in stem, as highlighted by the low translocation factor (Tf) measured in both clones. Net photosynthesis measurement, chlorophyll fluorescence images, transpiration values and chlorophyll content revealed a cadmium-induced impairment of physiological processes at the leaf level, which was more evident in clone Viglio. Metal binding and antioxidative compound content was differentially affected by cadmium exposure in the two eucalypt clones. Particularly, the content of thiols like cysteine and glutathione, organic acids like oxalate and citrate, and polyamines were markedly modulated in plant organs by metal treatment and highlighted different defence responses between the clones. Cadmium tolerance and accumulation ability of the eucalypt clones were evaluated and the potential of E. camaldulensis for the reclamation of metal polluted-waters is discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,4-Diaminobutane, 99%
Sigma-Aldrich
11-(1H-pyrrol-1-yl)undecane-1-thiol, 96%
Sigma-Aldrich
Spermine, ≥99.0% (GC)
Sigma-Aldrich
Spermine, ≥97%
Sigma-Aldrich
Spermine, suitable for cell culture, BioReagent
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Spermidine, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Spermidine, BioReagent, Molecular Biology, suitable for cell culture, ≥98%