Merck
Physical performance and life quality in postmenopausal women supplemented with vitamin D: a two-year prospective study.
Acta pharmacologica Sinica (2015-08-19)
Li-hong Gao, Wen-jun Zhu, Yu-juan Liu, Jie-mei Gu, Zhen-lin Zhang, Ou Wang, Xiao-ping Xing, Ling Xu
ABSTRAKT
To investigate the effects of calcium and vitamin D supplementation on bone turnover marker levels, muscle strength and quality of life in postmenopausal Chinese women. A total of 485 healthy postmenopausal Chinese women (63.44±5.04 years) were enrolled in this open-label, 2-year, prospective, community-based trial. The participants were divided into group A, B, C, which were treated with calcium (600 mg/d) alone, calcium (600 mg/d) and cholecalciferol (800 IU/d) or calcium (600 mg/d) and calcitriol (0.25 μg/d), respectively, for 2 years. Serum levels of 25-hydroxyvitamin D, parathyroid hormone, β-CTX and P1NP were measured, and the muscle strength and quality of life were assessed at baseline and at 12- and 24-month follow-ups. Four hundred and sixty one participants completed this study. Serum levels of 25-hydroxyvitamin D were significantly increased in group C, but not changed in groups A and B at 24-month follow-up. Serum levels of parathyroid hormone, bone turnover marker β-CTX and bone formation marker P1NP were significantly decreased in group C, while serum levels of β-CTX were increased in group A at 24-month follow-up. The participants in group C maintained the grip strength, while those in groups A and B exhibited decreased grip strength at 24-month follow-up. The quality of life for the participants in groups B and C remained consistent, but that in group A was deteriorated at 24-month follow-up. Supplementation with calcitriol and calcium modifies the bone turnover marker levels, and maintains muscle strength and quality of life in postmenopausal Chinese women, whereas supplementation with cholecalciferol and calcium prevents aging-mediated deterioration in quality of life.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, analytical standard