Merck
Effects of dietary cellulase and xylanase addition on digestion, rumen fermentation and methane emission in growing goats.
Archives of animal nutrition (2015-05-13)
Qi Lu, Jinzhen Jiao, Shaoxun Tang, Zhixiong He, Chuanshe Zhou, Xuefeng Han, Min Wang, Jinhe Kang, N E Odongo, Zhiliang Tan
ABSTRAKT
The objective of this study was to evaluate the effectiveness of supplementation of cellulase and xylanase to diets of growing goats to improve nutrient digestibility, utilisation of energy and mitigation of enteric methane emissions. The experiment was conducted in a 5 × 5 Latin square design using five goats with permanent rumen fistulae and five treatments consisted of two levels of cellulase crossed over with two levels of xylanase plus unsupplemented Control. The cellulase (243 U/g) derived from Neocallimastix patriciarum was added at 0.8 and 1.6 g/kg dry matter intake (DMI) and the xylanase (31,457 U/ml) derived from Aspergillus oryzae was fed at 1.4 and 2.2 ml/kg DMI. There were no differences in apparent digestibility of organic matter, neutral detergent fibre, acid detergent fibre and rumen fermentation parameters (i.e. ammonia-nitrogen [N], volatile fatty acids) among all treatments. Dietary cellulase and xylanase addition did not influence energy and N utilisation. But compared to xylanase addition at the higher dose, at the low xylanase dose the retained N, the availability of retained N and digested N were increased (p < 0.01). Moreover, enzyme addition did not affect the enteric methane emission and community diversity of ruminal methanogens. The present results indicated that previous in vitro findings were not confirmed in ruminant trials.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.99 atom % 12C
Węgiel - Szklisty, rod, 200mm, diameter 5.0mm, glassy carbon
Węgiel - Szklisty, foil, 50x50mm, thickness 1.0mm, glassy carbon
Węgiel - Szklisty, rod, 100mm, diameter 7.0mm, glassy carbon
Węgiel - Szklisty, foil, 50x50mm, thickness 4.0mm, glassy carbon
Węgiel - Szklisty, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
Węgiel - Szklisty, foil, 100x100mm, thickness 2.0mm, glassy carbon
Węgiel - Szklisty, rod, 200mm, diameter 1.0mm, glassy carbon
Węgiel - Szklisty, rod, 50mm, diameter 1.0mm, glassy carbon
Węgiel - Szklisty, foil, 100x100mm, thickness 6.0mm, glassy carbon
Węgiel - Szklisty, foil, 100x100mm, thickness 1.0mm, glassy carbon
Węgiel - Szklisty, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Węgiel - Szklisty, rod, 200mm, diameter 3.0mm, glassy carbon
Węgiel - Szklisty, foil, 10x10mm, thickness 1.0mm, glassy carbon
Węgiel - Szklisty, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Węgiel - Szklisty, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Węgiel - Szklisty, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Węgiel - Szklisty, rod, 100mm, diameter 5.0mm, glassy carbon
Węgiel - Szklisty, rod, 100mm, diameter 1.0mm, glassy carbon
Węgiel - Szklisty, foil, 25x25mm, thickness 0.5mm, glassy carbon
Węgiel - Szklisty, foil, 10x10mm, thickness 4.0mm, glassy carbon
Węgiel - Szklisty, rod, 100mm, diameter 3.0mm, glassy carbon
Węgiel - Szklisty, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Węgiel - Szklisty, foil, 25x25mm, thickness 4.0mm, glassy carbon
Węgiel - Szklisty, rod, 5mm, diameter 3.0mm, glassy carbon