Merck
Association of adipokines and inflammatory markers with lipid control in type 2 diabetes.
Polskie Archiwum Medycyny Wewnetrznej (2015-05-16)
Agnieszka Kapłon-Cieślicka, Marek Postuła, Marek Rosiak, Michał Peller, Agnieszka Kondracka, Agnieszka Serafin, Ewa Trzepla, Grzegorz Opolski, Krzysztof J Filipiak
ABSTRAKT
Data regarding the effect of certain adipokines on lipid metabolism are equivocal. The aim of this study was to evaluate the association of lipid control with adipokines and inflammatory markers in patients with type 2 diabetes. The analysis included 195 patients with type 2 diabetes. The achievement of treatment targets in terms of total cholesterol, low-density lipoprotein (LDL) cholesterol, high-density lipoprotein (HDL) cholesterol, and triglycerides was assessed in accordance with the current guidelines. Homeostatic Model Assessment-Insulin Resistance (HOMA-IR) index as well as concentrations of highmolecular-weight (HMW) adiponectin, leptin, resistin, high-sensitivity C-reactive protein, interleukin 6, and tumor necrosis factor α (TNF-α) were measured in all patients. Logistic regression analyses were performed to determine the risk factors for inadequate lipid control. Optimal control in terms of total cholesterol, LDL, HDL, and triglycerides was achieved in 61%, 43%, 53%, and 68% of the patients, respectively. In multivariate analyses, female sex, lower resistin concentrations, and the absence of statin treatment were predictors of total cholesterol levels above the treatment target; older age and lower statin dose--of LDL cholesterol levels above the treatment targets; female sex, higher HOMA-IR index, lower HMW adiponectin concentrations, and higher TNF-α concentration-o-f HDL levels below the treatment targets; and higher HOMA-IR, lower HMW adiponectin concentration, and the absence of statin treatment--of triglycerides above the treatment target. In type 2 diabetes, lower HMW adiponectin concentrations are associated with inadequate triglyceride and HDL control; higher TNF-α, with inadequate HDL control, and lower resistin concentrations, with inadequate total cholesterol control.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Aspirin, meets USP testing specifications