Merck
Au-Ag core-shell nanoparticle array by block copolymer lithography for synergistic broadband plasmonic properties.
ACS nano (2015-04-22)
Seung Keun Cha, Jeong Ho Mun, Taeyong Chang, Sang Yun Kim, Ju Young Kim, Hyeong Min Jin, Jeong Yong Lee, Jonghwa Shin, Kwang Ho Kim, Sang Ouk Kim
ABSTRAKT
Localized surface plasmon resonance of metallic nanostructures receives noticeable attention in photonics, electronics, catalysis, and so on. Core-shell nanostructures are particularly attractive due to the versatile tunability of plasmonic properties along with the independent control of core size, shell thickness, and corresponding chemical composition, but they commonly suffer from difficult synthetic procedures. We present a reliable and controllable route to a highly ordered uniform Au@Ag core-shell nanoparticle array via block copolymer lithography and subsequent seeded-shell growth. Size-tunable monodisperse Au nanodot arrays are generated by block copolymer self-assembly and are used as seed layers to grow Ag shells with variable thickness. The resultant Au@Ag core-shell nanoparticle arrays exhibit widely tunable broadband enhancement of plasmonic resonance, greatly surpassing single-element nanoparticle or homogeneous alloy nanoparticle arrays. Surface-enhanced Raman scattering of the core-shell nanoparticle arrays showed an enhancement factor greater than 270 from Au nanoparticle arrays.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%