Merck
Myelin basic protein as a novel genetic risk factor in rheumatoid arthritis--a genome-wide study combined with immunological analyses.
PloS one (2011-06-16)
Chikashi Terao, Koichiro Ohmura, Masaki Katayama, Meiko Takahashi, Miki Kokubo, Gora Diop, Yoshinobu Toda, Natsuki Yamamoto, Reiko Shinkura, Masakazu Shimizu, Ivo Gut, Simon Heath, Inga Melchers, Toshiaki Manabe, Mark Lathrop, Tsuneyo Mimori, Ryo Yamada, Fumihiko Matsuda
ABSTRAKT
Rheumatoid arthritis (RA) is a major cause of adult chronic inflammatory arthritis and a typical complex trait. Although several genetic determinants have been identified, they account for only a part of the genetic susceptibility. We conducted a genome-wide association study of RA in Japanese using 225,079 SNPs genotyped in 990 cases and 1,236 controls from two independent collections (658 cases and 934 controls in collection1; 332 cases and 302 controls in collection2), followed by replication studies in two additional collections (874 cases and 855 controls in collection3; 1,264 cases and 948 controls in collection4). SNPs showing p<0.005 in the first two collections and p<10(-4) by meta-analysis were further genotyped in the latter two collections. A novel risk variant, rs2000811, in intron2 of the myelin basic protein (MBP) at chromosome 18q23 showed strong association with RA (p = 2.7×10(-8), OR 1.23, 95% CI: 1.14-1.32). The transcription of MBP was significantly elevated with the risk allele compared to the alternative allele (p<0.001). We also established by immunohistochemistry that MBP was expressed in the synovial lining layer of RA patients, the main target of inflammation in the disease. Circulating autoantibody against MBP derived from human brain was quantified by ELISA between patients with RA, other connective tissue diseases and healthy controls. As a result, the titer of anti-MBP antibody was markedly higher in plasma of RA patients compared to healthy controls (p<0.001) and patients with other connective tissue disorders (p<0.001). ELISA experiment using citrullinated recombinant MBP revealed that a large fraction of anti-MBP antibody in RA patients recognized citrullinated MBP. This is the first report of a genetic study in RA implicating MBP as a potential autoantigen and its involvement in pathogenesis of the disease.