Merck
Elevated matrix metalloproteinase-7 expression promotes metastasis in human lung carcinoma.
World journal of surgical oncology (2015-01-16)
Ji-Chang Han, Xian-Dong Li, Jin Du, Feng Xu, Yu-Ju Wei, Hong-Bing Li, Yi-Jie Zhang
ABSTRAKT
Matrix metalloproteinase 7 (MMP-7) promotes tumor invasion and metastasis in several cancers. However, its role in lung cancer progression is understudied. In this study, we investigated the correlation between MMP-7 expression and lung cancer pathology. We searched the databases PubMed, Embase, Web of Science, Cochrane Library, CISCOM, CINAHL, China BioMedicine (CBM) and China National Knowledge Infrastructure (CNKI) for scientific literature relevant to MMP-7 and lung cancer. Carefully selected studies were pooled and ORs with 95% CI were calculated. Subgroup analyses and publication bias were analyzed to understand the retrieved data in greater detail. Version 12.0 STATA software was used for statistical analysis. We retrieved a total of 121 studies through database searches. Finally, 14 cohort studies satisfied our inclusion/exclusion criteria, and these 14 studies, published between 2004 and 2012, were selected for meta-analysis to understand the influence of MMP-7 expression in lung cancer progression. Our results showed consistent differences in MMP-7 expression when comparisons were made between TNM I-II versus III-IV (OR = 1.82, 95% CI: 1.19 to 2.78, P = 0.006); histologic grade 1 to 2 versus 3 to 4 (OR = 1.67, 95% CI: 1.14 to 2.42, P = 0.008); and lymph node-negative versus lymph node-positive samples (OR = 2.81, 95% CI: 1.73 to 4.58, P <0.001), with significantly higher MMP-7 expression levels found in the more advanced stages. Subgroup analysis showed that age was not the factor influencing the associations between histologic grade, LN metastasis and MMP-7 expression in lung cancer patients, as both under 60 and over 60 age groups showed strong correlations (all P <0.05). However, when TNM staging was analyzed for its association with MMP-7 expression, only patients under age 60 showed a statistically significant correlation. Our meta-analysis results revealed that MMP-7 overexpression is associated with advanced TNM and histological grades, and is linked to aggressive LN metastasis in lung cancer patients; thus MMP-7 is a useful biomarker to assess the disease status in lung cancers.