Merck
Clinical Significance of Wnt/β-Catenin Signalling and Androgen Receptor Expression in Prostate Cancer.
The world journal of men's health (2013-05-10)
Soo Jin Jung, Sangtaek Oh, Geun Taek Lee, Jaeil Chung, Kweonsik Min, Jangho Yoon, Wansuk Kim, Dong Soo Ryu, Isaac Yi Kim, Dong Il Kang
ABSTRAKT
To investigate the relationships among the Wnt/β-catenin pathway, androgen receptor (AR), and clinicopathological factors in hormone-naïve prostate cancer. This study was conducted with132 cases of hormone-naïve prostate cancer treated by prostatectomy and prostate needle biopsy. An immunohistochemical study using antibodies against β-catenin, matrix metalloproteinase-7 (MMP-7), and the AR was performed. For the in vitro study, PC-3, LNCaP, 22Rv1, and DU145 cell lines were used. The clinical or pathological stage ware a localized cancer in 36 patients (27.3%), locally advanced cancer in 31 (23.5%), and metastatic cancer in 65 (49.2%). We detected increased β-catenin, AR, and MMP-7 expression with a high Gleason grade, disease progression, and increasing serum prostate-specific antigen (PSA) levels (p<0.01). In Spearman's rank correlations, the expression of cytoplasmic β-catenin, MMP-7, and the AR were found to be significantly positively correlated. In addition, the expression of β-catenin, MMP-7, and the AR were significantly correlated with clinicopathological variables indicative of a poor prognosis. Forty-nine patients with primary androgen deprivation had short response durations from hormone therapy to PSA progression with elevated MMP-7 expression on the Kaplan-Meier curve (p=0.0036). These data show that an activated Wnt/β-catenin pathway and AR expression in prostate cancer are correlated with metastasis and aggressiveness. In addition, the expression of MMP-7 protein, a target of the Wnt/β-catenin pathway, is associated with PSA progression in prostate cancer patients undergoing primary hormone therapy.