Merck
  • Bioinformatics and reanalysis of subtracted expressed sequence tags from the human ciliary body: Identification of novel biological functions.
Bioinformatics and reanalysis of subtracted expressed sequence tags from the human ciliary body: Identification of novel biological functions.
Molecular vision (2002-10-02)
Julio Escribano, Miguel Coca-Prados
ABSTRAKT
The ciliary body is largely known for its major roles in the regulation of aqueous humor secretion, intraocular pressure, and accommodation of the lens. In this review article we applied bioinformatics to re-examine hundreds of expressed sequence tags (ESTs) previously isolated by subtractive hybridization from a human ciliary body library [1]. The DNA sequences of these clones have been recently added to the web site of NEIBank. DNA sequence comparisons of subtracted ESTs were performed against all entries in the last available release of the non-redundant database containing GenBank, EMBL, DDBJ and PDB sequences using the BlastN program accessed through NCBI's BLAST services on the internet (NCBI). Sequences were also compared and mapped using the Blast search program provided through the Internet by the Human Genome Project (UCSC). A total number of 284 independent ESTs were classified in 17 functional groups. Analysis of their relationships allowed to define the expression of five major groups of known genes: (i) protein synthesis, folding, secretion and degradation (20%); (ii) energy supply and biosynthesis (12%); (iii) contractility and cytoskeleton structure (6%); (iv) cellular signaling and cell cycle regulation (7%); and (v) nerve cell related tasks (2%), including neuropeptide processing and putative non-visual phototransduction and circadian rhythm control. The largest group contain unidentified sequences, a total of 105 sequences, accounting for 37% of ESTs. The unidentified sequences show similarity to genomic non-coding regions, or genes of unknown function. The most highly represented EST, correspond to myocilin, a gene involved in glaucoma. The data also confirms the secretory functions of the ciliary epithelium, and its high metabolism; the presence of a neuroendocrine peptidergic system presumably involved in the regulation of the intraocular pressure and/or aqueous humor secretion. Additional genes may be related to a non-visual phototransduction cascade and/or to circadian rhythms. Overall this initial group of subtracted ESTs can lead to uncover novel physiological functions of the ciliary body in normal and in disease, as well as novel candidate genes for ocular diseases.