Merck
Association of gene polymorphisms with chronic kidney disease in Japanese individuals.
International journal of molecular medicine (2009-09-03)
Tetsuro Yoshida, Kimihiko Kato, Kiyoshi Yokoi, Mitsutoshi Oguri, Sachiro Watanabe, Norifumi Metoki, Hidemi Yoshida, Kei Satoh, Yukitoshi Aoyagi, Yoshinori Nozawa, Yoshiji Yamada
ABSTRAKT
Chronic kidney disease (CKD) is recognized as a risk factor not only for end-stage renal disease but also for cardiovascular disease. Early detection and treatment of CKD is a likely key factor for prevention of its complications. Although genetic linkage analyses and association studies have implicated several loci and candidate genes in predisposition to CKD, the genes that underlie genetic susceptibility to this condition have remained largely unknown. The purpose of the present study was to identify genetic variants that confer susceptibility to CKD in Japanese individuals. The study population comprised 4,829 Japanese individuals (2,697 men, 2,132 women), including 757 subjects with CKD [464 men, 293 women; estimated glomerular filtration rate (eGFR) <50 ml min 1.73 m(-2)] and 4,072 controls (2,233 men, 1,839 women; eGFR >or=60 ml min 1.73 m(-2)). The genotypes for 40 polymorphisms of 39 candidate genes were determined. The chi-square test, multivariable logistic regression analysis with adjustment for covariates, as well as a stepwise forward selection procedure revealed that six polymorphisms of F10, PITRM1, PCSK2, JPH3, MYO7B, and AKAP12 were related (P<0.05) to the prevalence of CKD. Among these polymorphisms, the Cright curved arrow T polymorphism of F10 (rs5962) was most significantly associated with this condition. Determination of genotypes for the Cright curved arrow T polymorphism of F10 may prove informative for assessment of genetic risk for CKD in Japanese individuals.