Merck
PIR2/Rnf144B regulates epithelial homeostasis by mediating degradation of p21WAF1 and p63.
Oncogene (2012-11-07)
F Conforti, Ai Li Yang, M Cristina Piro, M Mellone, A Terrinoni, E Candi, P Tucci, G J Thomas, R A Knight, G Melino, B S Sayan
ABSTRAKT
ΔNp63 is a transcription factor that is critical for the development of stratified epithelia and is overexpressed or amplified in >80% of squamous cell carcinomas (SCCs). We identified the RING finger E3 ubiquitin ligase PIR2/Rnf144b as a direct transcriptional target of ΔNp63α and showed that its expression parallels that of ΔNp63α in keratinocytes, SCC cell lines and SCCs. We used primary keratinocytes as a model system to investigate the function of PIR2/Rnf144b in stratified epithelia. Depletion of PIR2/Rnf144b severely impaired keratinocyte proliferation and differentiation, associated with accumulation of p21(WAF1/CIP1); a known target of PIR2/Rnf144b. More importantly, we found that PIR2/Rnf144b binds and mediates proteasomal degradation of ΔNp63α, generating a hitherto unknown auto-regulatory feedback loop. These findings substantiate PIR2/Rnf144b as a potentially critical component of epithelial homeostasis, acting downstream of ΔNp63α to regulate cellular levels of p21(WAF1/CIP1) and ΔNp63α.