Merck
Effects of PPARγ agonistrosiglitazone on human retinoblastoma cell in vitro and in vivo.
International journal of clinical and experimental pathology (2016-01-02)
Xianyong Cao, Lin He, Yanhua Li
ABSTRAKT
The aim of the study was to evaluate the antitumor effects of the PPARγ agonist rosiglitazone on the human retinoblastoma. The cell biological behavior was detected, specifically, the effects of rosiglitazone on cell viability and apoptosis of the human retinoblastoma Y79 cells were investigated by MTT assay and Hochest 33258 staining and the migration assay showed that rosiglitazone blocked the invasion and migration of the carcinoma cells through the reconstituted extracellular matrix (Matrigel). The effect of rosiglitazone on NF-κB-dependent reporter gene transcription induced by LPS was analyzed by NF-κB-luciferase assay. Then human retinoblastoma Y79 cells were subcutaneously transplanted in BALB/c nude mice, and the animals were treated with rosiglitazone (20 mg/kg, 40 mg/kg, and 80 mg/kg) to verify its anti-tumor effect in vivo. Rosiglitazone suppressed the viability of Y79 cells dose- and time-dependently and induced apoptosis in Y79 cells in vitro. Molecular biology analysis found that rosiglitazone could modulate the proliferative and apoptosis related signal, reduce NF-κB-dependent reporter gene transcription induced by LPS. Rosiglitazone markedly reduced the growth of Y79 cells transplanted into the mice without causing significant side effects. Our results suggested that rosiglitazone demonstrated antitumor activity against the human retinoblastoma Y79 cells by inhibiting cell growth, inducing apoptosis and inhibiting metastasis and invasion in vitro and delaying tumor growth in vivo.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)